Learn More
Selleck Chemical LLC EI1 5mg 1418308-27-6
SDP
Supplier:  Selleck Chemical LLC S76115MG
Encompass_Preferred
EI1 is a potent and selective EZH2 inhibitor with IC50 of 15 nM and 13 nM for EZH2 (WT) and EZH2 (Y641F), respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5024