Learn More
Medchemexpress LLC Eicosanedioic acid | 2424-92-2 | 99.0% | 342.51 g/mol | C20H38O4 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0345955G
Encompass_Preferred
Eicosanedioic acid is a dicarboxylic acid used as a non-cleavable ADC linker and an alkyl-chain PROTAC linker for bioconjugation and linker chemistry research. It is supplied at high purity for use in synthesis of antibody-drug conjugates and targeted protein degradation constructs.
  • Dicarboxylic acid useful as a non-cleavable ADC linker.
  • Applicable as an alkyl-chain PROTAC linker in PROTAC synthesis.
  • High purity suitable for research use (99.0%).
  • Molecular weight 342.51 g/mol and formula C20H38O4.
  • Available in common lab pack sizes (5 G, 10 G, 25 G, 100 G).
Catalog No. 50-002-03192