Learn More
Medchemexpress LLC Eicosanedioic acid | 2424-92-2 | MFCD00059627 | 99.0% | 342.51 g/mol | C20H38O4 | 10 G

Supplier: Medchemexpress LLC HYW03459510G
Eicosanedioic acid (CAS 2424-92-2) is a 20-carbon dicarboxylic acid used as a non-cleavable alkyl linker in antibody-drug conjugate (ADC) and PROTAC synthesis and as a building block in organic synthesis. It is supplied at high purity for research applications.
- High purity (99.0%) suitable for research applications.
- Used as a non-cleavable ADC and PROTAC linker.
- Chemical formula C20H38O4 and molecular weight 342.51 g/mol.
- Available in laboratory pack sizes, including 10 G.
- Store sealed at 4°C and protect from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.