Learn More
Cayman Chemical EIcosapentaeNIc ACD mthyl 10mg
SDP
Supplier:  Cayman Chemical 900029510 MG
Encompass_Preferred
An esterified form of EPA; has been used as a reference standard in the quantification of fatty acids in microalgal and fish oils
Catalog No. 50-289-5706