Learn More
Cayman Chemical EIcosapentaenoIc ACD 1 2 3 4
SDP
Supplier:  Cayman Chemical 30017500 UG
Encompass_Preferred
An internal standard for the quantification of EPA by GC- or LC-MS
Catalog No. 50-298-9765