Learn More
Cayman Chemical EIcosapentaenoIc ACDd5 meth
SDP
Supplier:  Cayman Chemical 31126100 UG
Encompass_Preferred
An internal standard for the quantification of EPA methyl ester by GC- or LC-MS
Catalog No. 50-298-8609