Learn More
Cayman Chemical Eicosapentaenoyl Ethanolamide
SDP
Supplier:  Cayman Chemical 90018351 MG
Encompass_Preferred
An internal standard for the quantification of EPEA by GC- or LC-MS
Catalog No. 50-289-4244