Learn More
Cayman Chemical EIcosapentaenoyl EthnolamID
SDP
Supplier:  Cayman Chemical 9001835500 UG
Encompass_Preferred
An internal standard for the quantification of EPEA by GC- or LC-MS
Catalog No. 50-289-2542