Learn More
Medchemexpress LLC HY-125033 10mg , EIDD-1931 CAS:3258-02-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12503310MG
Encompass_Preferred
HY-125033 10mg EIDD-1931 CAS: 3258-02-4 EIDD-1931 (Beta-d-N4-hydroxycytidine; NHC) is a novel nucleoside analog and behaves as a potent anti-virus agent. EIDD-1931 effectively inhibits the replication activity of venezuelan equine encephalitis virus (VEEV), Chikungunya virus (CHIKV) and hepatitis C virus (HCV). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9177