Learn More
ABclonal Technology EIF2AK1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A141191000UL
Encompass_Preferred
The protein encoded by this gene acts at the level of translation initiation to downregulate protein synthesis in response to stress. The encoded protein is a kinase that can be inactivated by hemin. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-08410