Learn More
Sino Biological Peptide Substrates: EIF2S Peptide
Supplier:  Sino Biological E2358
Encompass_Preferred
The 13 amino acids of EIF2S peptide sequence (CILLSELSRRRIR) is derived from human protein EIF2S1 between residues 46-57. This peptide is suitable for use as a substrate for MNK1 and EIF2AK family kinases.
Catalog No. 50-256-9260