Learn More
Selleck Chemical LLC Elacestrant S9629-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S962910MM/1ML
Encompass_Preferred
Elacestrant (RAD1901) Dihydrochloride is an orally available selective estrogen receptor degrader (SERD) with IC50 of 48 nM and 870 nM for ER and ER respectively
Catalog No. 50-000-24937