Learn More
TARGETMOL CHEMICALS INC Elacestrant dihydrochloride
SDP
Supplier:  TARGETMOL CHEMICALS INC T136751MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Elacestrant dihydrochloride (DA-DKRAD1901 dihydrochloride) is an orally available selective estrogen receptor degrader with IC50s of 48 for ERalpha and 870 nM for ERbeta, respectively. Purity 98.46%
Catalog No. 50-287-5843