Learn More
Medchemexpress LLC Elacridar | 143664-11-3 | 99.7% | C34H33N3O5 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY50879200MG
Encompass_Preferred
Elacridar is an orally active inhibitor of P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP). It is utilized to study the impact of efflux transporters on agent distribution to the brain and in cancer research.
  • Examines the influence of efflux transporters on agent distribution to the brain.
  • Used in cancer research.
  • Inhibits cell viability in 786-O cells.
  • Affects P-glycoprotein and ABCG2 protein expression levels in MCF-7 and 786-O cell lines.
  • Increases intracellular accumulation in MCF-7 and 786-O cells.
Catalog No. 50-003-57804