Learn More
Medchemexpress LLC Elacridar | 143664-11-3 | MFCD00912604 | 97.5% | 563.64 | C34H33N3O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY50879R10MG
Encompass_Preferred
Elacridar is an orally active dual P-glycoprotein (P-gp) and breast cancer resistance protein (BCRP) inhibitor. This product is supplied as an analytical reference standard intended for research and analytical applications, with a reported purity of 97.5% and molecular formula C34H33N3O5.
  • Analytical reference standard for elacridar
  • Intended for research and analytical applications
  • Dual inhibitor of P-gp and BCRP for transporter studies
  • Reported purity 97.5%
  • Molecular formula C34H33N3O5; molecular weight 563.64 Da
  • Packaged as a 10 mg unit suitable for analytical assays
Catalog No. 50-002-98622