Learn More
Medchemexpress LLC HY-50879 50mg , Elacridar CAS:143664-11-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5087950MG
Encompass_Preferred
HY-50879 50mg Elacridar CAS: 143664-11-3 Elacridar is an orally active P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP) inhibitor. Elacridar can be used to examine the influence of efflux transporters on drug distribution to brain and the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0256