Learn More
Medchemexpress LLC Elacridar (GF120918) | 143664-11-3 | MFCD00912604 | 99.7% | C34H33N3O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY50879100MG
Encompass_Preferred
Elacridar is an orally active P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP) inhibitor. It can be utilized to investigate the influence of efflux transporters on agent distribution to the brain and in cancer research. This product is for research use only.
  • Inhibits P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP)
  • Utilized to examine efflux transporters on agent distribution to the brain
  • Used in cancer research
  • Affects P-glycoprotein and ABCG2 protein expression levels
  • Impacts 99mTc-MIBI intracellular accumulation
  • Shows differential distribution in brain and plasma
Catalog No. 50-003-57825