Learn More
Medchemexpress LLC Elacridar hydrochloride | 143851-98-3 | >98.0% | 600.10 | C34H34ClN3O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY5088010MG
Encompass_Preferred
Elacridar hydrochloride is an orally active inhibitor of P-glycoprotein (P-gp) and breast cancer resistance protein (BCRP), used in research to study efflux transporter-mediated drug distribution, including blood-brain barrier penetration and multidrug resistance in cancer. Supplied as the hydrochloride salt for research applications.
  • Orally active P-gp and BCRP inhibitor
  • Used to evaluate efflux transporter effects on drug distribution and blood-brain barrier penetration
  • Provided as hydrochloride salt for improved handling
  • Available in small research sizes for analytical and preclinical studies
  • Documented solubility and storage conditions for reproducible results
Catalog No. 50-002-98723