Learn More
Medchemexpress LLC Elacridar (Standard) | 143664-11-3 | C34H33N3O5 | 100 MG

Supplier: Medchemexpress LLC HY50879R100MG
Elacridar is a chemical compound (Catalog No. HY-50879R) for laboratory research use, identified by CAS No. 143664-11-3 and chemical formula C34H33N3O5. It appears as a solid with a melting point of 216-218°C and is stable under recommended storage conditions.
- Solid appearance
- Melting point 216-218°C
- Boiling point 701.6°C at 760 mmHg
- Relative density 1.264 g/cm³
- Store at -20°C, sealed, away from light and moisture
- For longer storage in solvent, store at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.