Learn More
TARGETMOL CHEMICALS INC ELACYTARABINE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T150095MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Elacytarabine (M7594 0037) a lipid-conjugated derivative of the nucleoside analog cytarabine is an antineoplastic drug. It has cytotoxicity in solid tumors. purity: 97%
Catalog No. 50-258-1260