Learn More
Enzo Life Sciences Elafin (human), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX201240C100
Encompass_Preferred
Alternative name: ESI, Elastase specific inhibitor, SKALP, Skin derived antileukoproteinase, Protease inhibitor WAP3. Purity: ≥90% (HPLC, SDS-PAGE, MS). MW: ~5.9kDa. Formulation: Lyophilized. Source: Produced in yeast. Long Term Storage: -20°C.
Catalog No. 50-200-7955