Learn More
Medchemexpress LLC Elagolix sodium | 832720-36-2 | 99.9% | C32H29F5N3NaO5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1436910MM/1ML
Encompass_Preferred
Elagolix sodium is a highly effective, selective, oral-active, and short-term non-peptide gonadotropin-releasing hormone receptor (GnRH receptor) antagonist. It also acts as an NFAT inhibitor and effectively blocks Ca2+ flux. This compound is suitable for studying pain related to endometriosis and other related conditions.
  • Highly effective and selective GnRH receptor antagonist
  • Oral-active and short-term action
  • Acts as an NFAT inhibitor
  • Suitable for studying pain related to endometriosis
Catalog No. 50-003-54257