Learn More
Cayman Chemical ElaIdIc AcId-d17 5mg
SDP
Supplier:  Cayman Chemical 277155 MG
Encompass_Preferred
An internal standard for the quantification of elaidic acid by GC- or LC-MS
Catalog No. 50-294-1471