Learn More
Medchemexpress LLC Elaidic acid (Standard) | 112-79-8 | 99.8% | 282.46 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY113016R100MG
Encompass_Preferred
Elaidic acid is an analytical standard intended for research and analytical applications. It is the major trans fat found in hydrogenated vegetable oils and can be used as a pharmaceutical solvent.
  • Used in qualitative research experiments.
  • Used in quantitative research experiments.
  • Used in methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-39719