Learn More
STEMCELL Technologies Elastase, Size: 1 g
SDP
Supplier:  STEMCELL Technologies 1000682
Encompass
Elastase is a serine protease consisting of a single polypeptide chain of 240 amino acid residues and contains four disulfide bridges (Shotton Hartley). Elastase is synthesized as a zymogen (proelastase) and is converted to the active form by limited trypsin proteolysis at its N-terminal. Elastase preferentially cleaves the peptide bond of C-terminal neutral non-aromatic amino acid residues (Schellenberger et al.). Elastase is frequently used in combination with other proteases such as collagenase and trypsin to digest fibrous tissues because of its ability to digest native elastin.
Catalog No. 50-239-0121