Learn More
Chondrex Inc Elastase from Porcine Pancreas
Supplier:  Chondrex Inc 30047
Encompass_Preferred
Use to digest polymeric collagens from tissue samples in neutral buffer. Keep collagen samples at 4°C to avoid denaturation. PURITY: Purified by ion-exchange chromatography, ≥ 8 units/mg protein (E1%/280)
Catalog No. NC1929643