Learn More
Medchemexpress LLC Elastase-IN-3 25g
SDP
Supplier:  Medchemexpress LLC 25GHYW104758
Encompass_Preferred
Elastase-IN-3 (Compound 2i) is an Elastase inhibitor of Pseudomonas aeruginosa Elastase-IN-3 inhibits the quorum sensing system of Pseudomonas aeruginosa by targeting the LasR protein (KD value of 7 04 M) thereby preventing food spoilage caused by Pseudomonas aeruginosa[1]
Catalog No. 50-002-05452