Learn More
Abcam Elastatinal, elastase inhibitor, 25MG
Supplier:  Abcam AB14454125MG
Encompass_Preferred
MW 512.6 Da, Purity >98%. Selective, irreversible elastase inhibitor. Inhibits succinyltrialanine p-nitroanilide hydrolytic enzyme (IC₅₀ = 17 μM, porcine liver). Inhibits neutrophil and microglial elastase. Shows antiviral effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5642