Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Karcher Nozzle, PVC, DN40, ESD Safe

Supplier:  MSC 48041925

Encompass

This standard nozzle, constructed from durable Polyvinylchlorid (PVC), is engineered for optimal performance with industrial vacuum cleaners. It features a DN40 nominal width and is ESD safe, ensuring safe operation in environments requiring static discharge protection. With an overall length of 11.81 inches and a compatible hose diameter of 1.50 inches, this nozzle is designed for efficient and reliable cleaning.

  • Constructed from polyvinylchlorid (PVC).
  • Features a DN40 nominal width.
  • ESD safe.
  • Overall length of 11.81 inches.
  • Compatible with 1.50 inch hose diameter.
  • Packaged weight of 0.2 kg.

Catalog No. 50-283-7675


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.