Learn More
Cusabio Technology LLC Human Elastin,ELN ELISA Kit
Supplier:  Cusabio Technology LLC CSBE09338H
Encompass_Preferred
This Human ELN ELISA Kit was designed for the quantitative measurement of Human ELN protein in serum, plasma, cell culture supernates, tissue homogenates, cell lysates, urine. It is a Sandwich ELISA kit, its detection range is 3.12 pg/mL-200 pg/mL and the sensitivity is 0.78 pg/mL . One kit contains 96 wells.
Catalog No. NC2952930