Learn More
Medchemexpress LLC Elastin from bovine | 9007-58-3 | 25 MG

Supplier: Medchemexpress LLC 25MGHYP2841A
Elastin from bovine is a biochemical reagent intended for research use only. It is supplied as a solid, appearing off-white to light yellow.
- Intended for research use only
- Solid, off-white to light yellow appearance
- Recommended storage at -20°C, protected from light
- In solvent, recommended storage at -80°C for 6 months or -20°C for 1 month
- Soluble in H2O at concentrations of 10 mg/mL or greater
- Stable under recommended storage conditions
- Not classified as a hazardous substance or mixture
- Certificate of Analysis, Safety Data Sheet, and handling instructions are available
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.