Learn More
AVANTOR MASTERFLEX (VWR LLC) Parker Hannifin Compression Female Threaded Adapter Elbow, PFA, 1/2"" OD X 1/2"" NPT(F)
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3132184
Encompass
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Catalog No. 50-631-905