Learn More
AVANTOR MASTERFLEX (VWR LLC) ELBOWS 1/8FTFT X 3/8FTFT 10/P
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4061055
Encompass
Masterflex Fitting, Natural Polypropylene, Elbow, Hosebarb to Thread, 3/8IN ID x 1/8IN NPT(M), Avantor;10/PK, Avantor
Catalog No. 50-274-2356