Learn More
Med Vet International Eldridge Anesthesia Mask, J0355S, Set of 3
Supplier:  Med Vet International JOR355S
Encompass_Preferred
Set Of 3 The Eldridge Anesthesia Mask addresses a common problem. A standard round mask wants to roll away from patient on a flat table. Towels are often used to brace the mask into position. The Eldridge mask is flat on one side to prevent rolling ans sa
Catalog No. 50-005-82967