Learn More
MSC Kranzle Pressure Washer, Cold Water, Electric, 2.5 GPM, Heavy-Duty Axial Cam
Supplier:  MSC 37143419
Encompass
This cold water electric pressure washer is designed for industrial and commercial use, making it suitable for professionals in factories, food processing plants, vehicle wash bays, or farms for cleaning animal pens.
  • Advanced pump design offers multi-hour bypass and run-dry capability, preventing accidental pump damage.
  • Turbo nozzle doubles cleaning efficiency.
  • Unique high-pressure soap design delivers 20 percent more cleaning power at the tip.
  • Auto on/off control at the gun jet with a 30-second time delay reduces wear and tear on the pump.
  • TEFC 1600 RPM electric 220V, 15A, single phase motor with GFI.
Catalog No. 50-283-7721