Learn More
MSC PRO-SOURCE Pressure Washer, Electric, Cold Water, 3 GPM, 2000 PSI
Supplier:  MSC 48352272
Encompass
The PRO-SOURCE EC320M is an electric cold water pressure washer designed for industrial applications. It features a 3.0 GPM flow rate, 2000 PSI pressure, and is powered by a 220V electric motor with an AR Triplex Ceramic Plunger Pump, making it suitable for heavy-duty cleaning tasks.
  • Cold water operation
  • Electric powered
  • 3.0 GPM flow rate
  • 220V voltage
  • 20.0 A maximum amperage
  • Universal motor driven
  • AR Triplex Ceramic Plunger pump
  • Single phase power
  • 50.00 foot hose length
  • Integrated 2 wheel cart frame
  • 2000 PSI pressure
  • 3/8 inch hose size
Catalog No. 50-283-8963