Learn More
Grainger Insulating Electrical Tape: Gen Purpose, Super 33+, Vinyl, 3/4 in x 76 ft, Black
Supplier:  Grainger 2A225
Encompass_Preferred
Scotch Super 33+ premium-grade, all-weather electrical tape features an aggressive adhesive for a secure hold. The flexible vinyl construction conforms to irregular surfaces in temps. down to 0°F. Provides you a flame-retardant material with a pressure–sensitive, rubber-based adhesive compatible with solid dielectric cable insulations. Resistant to abrasion, acids, alkalies, corrosion, moisture, and UV exposure.
Catalog No. NC1311220