Learn More
Med Vet International Electorsurgical Unit Electrode: Small Oval Loop, J0470DT7
Supplier: Med Vet International JOR470DT7
Autoclavable. 1/16" Use only with JOR470 unitNOTE: DO NOT store in liquids.