Learn More
Ted Pella Inc ELECTRODAG 502 30G PAINT
SDP
Supplier:  Ted Pella Inc 16056
Encompass
Conductive carbon glue with an acrylic binder. The acrylic binder provides a much stronger bond than water or isopropanol based carbon paints. This carbon glue forms a thin, conductive and flexible layer with excellent adhesion on a wide variety of substrates. Ideally suited for conductive bonding or conductive painting where a strong bond and good conductivity is required. Let air dry or speed up heating up to max. 65°C (149°F). Supplied in a bottle with a brush, 30g. Acetone (#16023) can be used as thinner/extender or as a cleaner to remove the carbon glue. •Service Temperature: -40 to +120°C (-40 to +248°F) •Solid Contents (graphite and carbon black): 39% •Flake size: 5-10µm •Binder: Acrylic •Carrier: Acetone based •Sheet resistance: 47 ohm/sq
Catalog No. NC9454656