Learn More
Hach Company Hach Electrode Cleaning Solution RENOVO.N, 250 mL - For pH Electrode Maintenance
Supplier: Hach Company S16M001
Hach Electrode Cleaning Solution (RENOVO.N) is a gentle reagent formulated for routine maintenance of pH sensing elements used in clean water sampling. It provides soft cleaning to remove light surface contamination and helps maintain accurate electrode response between deeper cleanings.
- Gently cleans pH sensing elements to remove light surface contamination.
- 250 mL bottle suitable for routine maintenance.
- Suitable for use after measurements in clean water samples and common water treatment applications.
- Helps maintain accurate pH readings and prolong sensor life.
- Easy to apply; follow safety and handling instructions on the product label.