Learn More
Hach Company Sension Fluoride Ion Selective Electrode (ISE), BNC connector
Supplier:  Hach Company 5192800
Encompass
Combination fluoride ion selective electrode (ISE) with replaceable gel electrolyte and fixed cable with BNC connector. Probe comes with extra replacement electrolyte gel reference cartridge. Compatible with Hach Sension, Sension+ and H-Series systems.
Catalog No. 50-199-7147