Learn More
Hach Company Electrodeless Conduct Sens-San
Supplier:  Hach Company 3708E2T50N
Encompass
Electrodeless Conductivity Sensor-Sanitary Style-PFA Body Material-50-ft CablePFA Sanitary (CIP) Flange Mounting Style 15m (50 ft) Analog Cable Electrodeless Conductivity Sensor
Catalog No. 50-002-07534