Learn More
Grainger Electronic Timer 7 Days (2) SPST-NO
Supplier:  Grainger 14H247
Encompass_Preferred
The OMRON Electronic Timer 7-day is a programmable timer that can be set for up to 7 days. It has an LCD display that shows the current time, on/off status, and settings. The timer allows you to set up to 8 on/off programs per day with 1-minute increments. It has a manual override on/off switch and requires 2 AA batteries (not included). This electronic timer can be used to control lights, pumps, heaters and more up to 15A. It measures 2.4 x 4.5 x 1.4 inches.
Catalog No. 50-275-0878