Learn More
Medchemexpress LLC Eledoisin Related Peptide | 2990-43-4 | 99.55% | 706.94 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP11865MG
Encompass_Preferred
Eledoisin Related Peptide is a Substance P analog that excites neurons and triggers behavioral responses. It is also a tachykinin receptor ligand.
  • Excites neurons
  • Triggers behavioral responses
  • Tachykinin receptor ligand
  • Shares a common N-terminal amino acid sequence with Substance P (SP)
  • Exhibits SP-like activity in the periphery (gut and salivary glands) and the CNS
  • Approximately equipotent with Substance P at identical ejection currents on single-cell activity of neurons
  • Has excitatory effects on the activity of respiratory neurons and reflex interneurons, similar to glutamate and Substance P
Catalog No. 50-004-24521