Learn More
Medchemexpress LLC HY-N2454 5mg Medchemexpress, β-Elemonic acid CAS:28282-25-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN24545MG
Encompass_Preferred
Medchemexpress, HY-N2454 5mg β-Elemonic acid CAS:28282-25-9 β-Elemonic acid is a triterpene isolated from Boswellia papyrifera. β-Elemonic acid induces cell apoptosis, reactive oxygen species (ROS) and COX-2 expression and inhibits prolyl endopeptidase. β-Elemonic acid exhibits anticancer and anti-inflammatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2897