Learn More
Selleck Chemical LLC Elenestinib phosphate-100MG
SDP
Supplier:  Selleck Chemical LLC E1662100MG
Encompass_Preferred
Elenestinib phosphate(BLU-263 phosphate) is a potent and orally active inhibitor of tyrosine kinase that exhibits potent inhibition of c-KIT (D816V) mutation BLU-263 phosphate has the potential for its use in the research of systemic mastocytosis (SM)
Catalog No. 50-313-6028