Learn More
Apexbio Technology LLC Eletriptan HBr 177834-92-3 50mg
SDP
Supplier:  Apexbio Technology LLC B223750
Encompass_Preferred
Eletriptan hydrobromide (CAS 177834-92-3) is an orally bioavailable small molecule that acts as a selective agonist for the 5-hydroxytryptamine (serotonin) 5-HT1B and 5-HT1D receptors By activating these receptor subtypes eletriptan hydrobromide modulates cranial blood vessel tone and neuronal signaling implicated in migraine pathophysiology This compound is commonly utilized in preclinical and clinical research to investigate serotonergic modulation of neurovascular function and to explore therapeutic mechanisms relevant to migraine and related neurovascular disorders
Catalog No. 50-000-92069