Learn More
Selleck Chemical LLC Eletriptan HBr S3180-10mg
SDP
Supplier:  Selleck Chemical LLC S318010MG
Encompass_Preferred
Eletriptan (UK-116044) is a selective 5-HT1B and 5-HT1D receptor agonist with Ki of 0 92 nM and 3 14 nM respectively
Catalog No. 50-310-8568