Learn More
Medchemexpress LLC Eletriptan hydrobromide | 177834-92-3 | 99.8% | C22H27BrN2O2S | 1 ML
SDP
Supplier:  Medchemexpress LLC HYA001010MM/1ML
Encompass_Preferred
Eletriptan hydrobromide is a 5-HT1B and 5-HT1D receptor agonist with antimigraine activity, primarily used as a laboratory chemical for research purposes. It appears as an off-white to light brown solid with a molecular weight of 463.43.
  • 5-HT1B and 5-HT1D receptor agonist
  • Demonstrates antimigraine activity
  • Ki values of 0.92 nM for 5-HT1B receptor
  • Ki values of 3.14 nM for 5-HT1D receptor
  • Off-white to light brown solid appearance
  • Reduces carotid blood flow in anesthetized dogs (in vivo)
  • Inhibits dural plasma protein extravasation in anesthetized rats (in vivo)
Catalog No. 50-004-32451