Learn More
Selleck Chemical LLC Eleutherol E8270-5MG
SDP
Supplier:  Selleck Chemical LLC E82705MG
Encompass_Preferred
Eleutherol (Allium A) is a naphthalene extracted from the bulb of E americana and a potent inhibitor of -glucosidase with an IC50 of 1 00 M It also exhibits significant antifungal activity with MIC values between 7 8 g/mL and 250 g/mL It can be used in research on antimicrobial infections and diabetes treatment
Catalog No. 50-000-41922